Please login to your online account to display your discounted pricing

Alfa Aesar™ Lutetium(III) fluoride, REacton™, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Lutetium(III) fluoride
CAS 13760-81-1
Purity Grade Reacton™
Formula Weight 231.97
MDL Number MFCD00016191
UN Number UN3288
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1126103

  SDS
alfa aesar™
1126103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1126106

  SDS
alfa aesar™
1126106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Lumps
Formula Weight 231.97
MDL Number MFCD00016191
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99.99%
Melting Point 1182°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
Density 8.3g/mL
Odor Odorless
Health Hazard 1 Danger
UN Number UN3288
Quantity 1g
Chemical Name or Material Lutetium(III) fluoride
Sensitivity Hygroscopic
Assay Percent Notes (REO)
CAS 13760-81-1
TSCA TSCA
Purity Grade Reacton™
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.