Please login to your online account to display your discounted pricing

Alfa Aesar™ Lutetium(III) chloride hexahydrate, REacton™, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Lutetium(III) chloride hexahydrate
CAS 15230-79-2
Purity Grade Reacton™
Formula Weight 389.42 (281.33 Anhydrous)
MDL Number MFCD00011099
Merck Index 14,5615
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1126003

 
alfa aesar™
1126003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1126006

 
alfa aesar™
1126006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 389.42 (281.33 Anhydrous)
Chemical Name or Material Lutetium(III) chloride hexahydrate
TSCA TSCA
Purity Grade Reacton™
Physical Form Crystalline Aggregates
Assay 99.9%
Assay Percent Notes (REO)
MDL Number MFCD00011099
CAS 15230-79-2
Merck Index 14,5615
Quantity 1g