missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lutetium(III) chloride hexahydrate, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1126006

 View more versions of this product

Catalog No. AA1126006



Specifications

Specifications

Lutetium(III) chloride hexahydrate
(REO)
14,5615
Crystalline Aggregates
Reacton™
15230-79-2
MFCD00011099
389.42 (281.33 Anhydrous)
99.9%
5g

Chemical Identifiers

15230-79-2
MFCD00011099
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents