Please login to your online account to display your discounted pricing

Alfa Aesar™ Lutetium(III) chloride, anhydrous, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Lutetium(III) chloride
CAS 10099-66-8
Name Note Anhydrous
Formula Weight 281.33
MDL Number MFCD00011099
Merck Index 14,5615
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1868503

  SDS
alfa aesar™
1868503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1868506

  SDS
alfa aesar™
1868506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 281.33
Chemical Name or Material Lutetium(III) chloride
TSCA TSCA
Mesh Size −20 Mesh
Name Note Anhydrous
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
MDL Number MFCD00011099
CAS 10099-66-8
Merck Index 14,5615
Melting Point 905°C (subl.)
Quantity 1g
Density 3.98g/mL