missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lutetium(III) chloride, anhydrous, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1868506

 View more versions of this product

Catalog No. AA1868506



Specifications

Specifications

Lutetium(III) chloride
10099-66-8
MFCD00011099
281.33
3.98g/mL
Hygroscopic
5g
Anhydrous
(REO)
14,5615
99.9%
−20 Mesh
905°C (sublimation)

Chemical Identifiers

10099-66-8
MFCD00011099
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents