Please login to your online account to display your discounted pricing

Lutetium(III) Chloride, 99.9%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC383540010

 View more versions of this product

Catalog No. AC383540010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Purity Grade Trace Metal Basis
Formula Weight 281.33
Quantity 1g
Color White to light gray
Linear Formula LuCl3
Molecular Formula Cl3Lu
CAS 10099-66-8
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
MDL Number MFCD00011099
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Total Trace Metal Impurities 0.15% max.
Assay Percent Range 99.9%
Chemical Name or Material Lutetium(III) chloride

Specifications

Chemical Name or Material Lutetium(III) chloride
CAS 10099-66-8
Purity Grade Trace Metal Basis
Assay Percent Range 99.9%
Molecular Formula Cl3Lu
Linear Formula LuCl3
Formula Weight 281.33
MDL Number MFCD00011099
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents