Please login to your online account to display your discounted pricing

Lutetium(III) Chloride, 99.9%, ACROS Organics™

$692.90 - $692.90

Chemical Identifiers

Chemical Name or Material Lutetium(III) chloride
CAS 10099-66-8
Purity Grade Trace Metal Basis
Assay Percent Range 99.9%
Molecular Formula Cl3Lu
Linear Formula LuCl3
Formula Weight 281.33
MDL Number MFCD00011099
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383540050

  SDS
thermo fisher scientific
AC383540050
Glass bottle 5g Each for $692.90

AC383540010

  SDS
thermo fisher scientific
AC383540010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Trace Metal Basis
Physical Form Powder
Formula Weight 281.33
Quantity 5g
Color White to light gray
Linear Formula LuCl3
Chemical Name or Material Lutetium(III) chloride
Packaging Glass bottle
Molecular Formula Cl3Lu
MDL Number MFCD00011099
CAS 10099-66-8
Total Trace Metal Impurities 0.15% max.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Assay Percent Range 99.9%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For ultra trace analysis

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.