missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lutetium(III) acetate hydrate, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1457106

 View more versions of this product

Catalog No. AA1457106



Specifications

Specifications

Lutetium(III) acetate hydrate
(REO)
352.11 (Anhydrous)
99.9%
Hygroscopic
207500-05-8
MFCD00211310
Crystalline Aggregates
Reacton™
5g

Chemical Identifiers

207500-05-8
MFCD00211310
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents