missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lutetium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01318906
Description
Lutetium(III) 2,4-pentanedionate is involved in refluxing 1,2,4-trichlorobenzene. It is also used as catalyst of organic compound.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lutetium(III) 2,4-pentanedionate | |
Powder | |
(REO) | |
MFCD00078030 | |
Insoluble in water. | |
[Lu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
472.29 | |
472.3 | |
Reacton™ |
17966-84-6 | |
5 g | |
C15H21LuO6 | |
lutetium iii acetylacetonate hydrate, lutetium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy lutetio oxy pent-3-en-2-one | |
HXFHOORFYPCGNF-UHFFFAOYSA-N | |
(Z)-4-hydroxypent-3-en-2-one;lutetium | |
24282830 | |
99.9% | |
Hygroscopic |
Chemical Identifiers
17966-84-6 | |
472.29 | |
HXFHOORFYPCGNF-UHFFFAOYSA-N | |
24282830 |
C15H21LuO6 | |
MFCD00078030 | |
lutetium iii acetylacetonate hydrate, lutetium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy lutetio oxy pent-3-en-2-one | |
[Lu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only