Please login to your online account to display your discounted pricing

Alfa Aesar™ Lutetium foil, 0.127mm (0.005 in.) thick, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Lutetium foil
CAS 7439-94-3
Name Note 0.127mm (0.005 in.) thick
MDL Number MFCD00011098
UN Number UN3178
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA00599FF

  SDS
alfa aesar™
00599FF
25 x 25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA00599FI

  SDS
alfa aesar™
00599FI
50 x 50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25 x 25mm
Packaging Mylar under Argon
Chemical Name or Material Lutetium foil
Odor Odorless
TSCA TSCA
Name Note 0.127mm (0.005 in.) thick
Physical Form Foil
Assay 99.9%
Assay Percent Notes (REO)
MDL Number MFCD00011098
CAS 7439-94-3
UN Number UN3178
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H260-H228
In contact with water releases flammable gases which may ignite spontaneously.
Flammable solid.
Health Hazard 3 GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Packaged under argon
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
Weight ≈0.78g/25x25mm
Color Silver-gray

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H260-H228
In contact with water releases flammable gases which may ignite spontaneously.
Flammable solid.

  • Packaged under argon