missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lutetium, AAS standard solution, Lu 1000μg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 89886AE

 View more versions of this product

Catalog No. AA89886AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Lutetium
AAS Standard
MFCD00011098
Liquid
100mL
12032-20-1,7697-37-2,7732-18-5
Lu2O3 in 5% HNO3
Nitric acid: 5%; Lutetium(III) oxid: 0.11%; Water: 94.89%
17mm Hg at 20°C

Chemical Identifiers

12032-20-1
Lu2O3 in 5% HNO3
MFCD00011098
Safety and Handling

Safety and Handling

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-714-2

SDS Documents