Please login to your online account to display your discounted pricing

Lutetium, AAS standard solution, Lu 1000μg/mL, Alfa Aesar™, Specpure™

Chemical Identifiers

Chemical Name or Material Lutetium
CAS 7697-37-2
CAS 12032-20-1
CAS 7732-18-5
Purity Grade AAS Standard
Physical Form Liquid
Molecular Formula Lu2O3 in 5% HNO3
MDL Number MFCD00011098
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA89886AE

 
alfa aesar
89886AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA89886AP

 
alfa aesar
89886AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade AAS Standard
Quantity 100mL
DOT Information DOT Class: 8, Packaging Group: III
Chemical Name or Material Lutetium
EINECS Number 231-714-2
Physical Form Liquid
Molecular Formula Lu2O3 in 5% HNO3
MDL Number MFCD00011098
CAS 7697-37-2
Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Lutetium(III) oxid: 0.11%; Water: 94.89%
Health Hazard 1 H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.
CAS 12032-20-1
Vapor Pressure 17mm Hg at 20°C
CAS 7732-18-5
Safety and Handling

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.