missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Luteolin, Tocris

Anti-inflammatory, antioxidant and free radical scavenger

$112.75 - $445.93

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C15H10O6
Formula Weight 286.24
Synonym 2-(3, 4-Dihydroxyphenyl)-5, 7-dihydroxy-4H-1-benzopyran-4-one
Chemical Name or Material Luteolin
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
287410 R & D Systems
RDS287410
10mg Each for $112.75
287450 R & D Systems
RDS287450
50mg Each for $445.93
Specifications Description & Specifications

Chemical Identifiers

>98%
286.24
Luteolin
C15H10O6
2-(3, 4-Dihydroxyphenyl)-5, 7-dihydroxy-4H-1-benzopyran-4-one

Specifications

491-70-3
C15H10O6
2-(3, 4-Dihydroxyphenyl)-5, 7-dihydroxy-4H-1-benzopyran-4-one
Luteolin
>98%
286.24
10mg

Safety and Handling

Recommended Storage : Store at +4°C