Please login to your online account to display your discounted pricing

Alfa Aesar™ (-)-Lupinine, 97%

$81.22 - $81.22

Chemical Identifiers

Chemical Name or Material (-)-Lupinine
CAS 486-70-4
Molecular Formula C10H19NO
Formula Weight 169.27
MDL Number MFCD00213431
UN Number UN1544
Beilstein 80447
Merck Index 14,5609
Synonym (1R-trans)-Octahydro-2H-quinolizine-1-methanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL18485MC

  SDS
alfa aesar™
L18485MC
100mg Each for $81.22

AAL18485ME

  SDS
alfa aesar™
L18485ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.27
Recommended Storage 2° to 8°C
Assay 97%
MDL Number MFCD00213431
Chemical Name or Material (-)-Lupinine
Health Hazard 2 GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 486-70-4
Health Hazard 1 Warning
Synonym (1R-trans)-Octahydro-2H-quinolizine-1-methanol
Merck Index 14,5609
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
Beilstein 80447
Molecular Formula C10H19NO
Optical Rotation −26° (c=3 in Water)
UN Number UN1544
Quantity 100mg
Boiling Point 160° to 164°C (4mmHg)
Melting Point 62° to 65°C
RTECS Number OK5802000
Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

2° to 8°C