Please login to your online account to display your discounted pricing

American Educational Products Lung/Breath Volume Kits


Investigate pulmonary fitness.

Manufacturer: american educational products  6109

 View more versions of this product

Catalog No. S16206

  • / Each
 

Description & Specifications

Specifications

Age 8 to 15 years
For Use With (Application) To measure lung volume with reusable components
Grade Level 3 to 9
Includes Durable plastic bag, mouthpiece and complete instruction guide
Product Type Breath Volume Kit