missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

American Educational Products Lung/Breath Volume Kits

Investigate pulmonary fitness.

$33.80 - $61.45

Specifications

For Use With (Application) To measure lung volume with reusable components
Grade Level 3 to 9
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Product Type Price Quantity  
Catalog Number Mfr. No. Includes Product Type Price Quantity  
S16205 American Educational Products
6050EMD
Durable plastic bag calibrated to 6l, mouthpiece and mouthpiece holder Lung Volume Kit Each for $61.45
S16206 American Educational Products
6109
Durable plastic bag, mouthpiece and complete instruction guide Breath Volume Kit Each for $33.80
Specifications

Description

  • Lung Volume Kit allows you to measure lung volume with reusable components
  • Breath Volume Kit measures tidal and respiratory minute volume

Description & Specifications

Specifications

To measure lung volume with reusable components
3 to 9