missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Luminaris Color HiGreen qPCR Master Mix, high ROX
Thermo Scientific Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR.
Supplier: Thermo Scientific™ K0363
Includes:
- Luminaris Color HiGreen High ROX qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer with a blue dye. The master mix contains high concentration of ROX passive reference
Description
Thermo Scientific™ Luminaris Color HiGreen Master Mixes are ready-to-use qPCR master mixes with color and premixed UDG optimized for SYBR Green chemistry using standard cycling protocols.
- Blue master mix and Yellow Sample Buffer for easy pipetting
- UDG in the master mix to prevent carry-over contamination
- Equal performance of blue and colorless master mixes
- Versions with premixed ROX or fluorescein for different real-time platforms
- Standard cycling protocols
- Excellent qPCR performance
- Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
- Sensitivity: single copy detection
- Wide linear range: accurate quantification across 9 orders of magnitude
- Reproducibility and convenience: ready-to-use 2X master mix
Specifications
Contains: • 10 x 1.25 mL Luminaris Color HiGreen High ROX qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer with a blue dye; sufficient for 1250 x 20 μL reactions. The master mix contains high concentration of ROX passive reference dye for specific real-time platforms. • 3 x 1.25 mL 40X Yellow Sample Buffer • 10 x 1.25 mL Nuclease-Free Water Store at -20°C. |
|
High | |
Taq DNA Polymerase | |
7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System | |
qPCR Master Mix | |
Gene Expression | |
2 X | |
DNA, dsDNA |
SYBR | |
qPCR | |
Standard | |
Luminaris | |
250 Reactions | |
2X | |
1250 Reactions |
For Research Use Only. Not for use in diagnostic procedures.