missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
LumiLoupe™ Magnifiers
For insects, plants, maps and graphics.
Manufacturer: Fisher Science Education LL77
Description
- Stand magnifiers reveal extraordinary details in everyday objects
- Dual lenses allow close-up examinations of objects
- Transparent acrylic base permits ambient light to illuminate the subject for bright, clear viewing
10X Magnification
- Pre-focused lenses
- Dimensions: 2.0 × 2.0 × 2.0 in. (5.1 × 5.1 × 5.1cm)
- Adjustable focus
- Dimensions: 2.0 × 2.0 × 1.75 in. (5.1 × 5.1 × 4.4cm)
Specifications
7x | |
For viewing fine details, coins, stamps, photography, miniatures, insects, plants, maps, graphics | |
111.98g |
Lumiloupe™ Stand Magnifier | |
3.95 oz. |