Please login to your online account to display your discounted pricing

LUF 6283, Tocris™

HCA2 (GPR109A) partial agonist

$129.05 - $546.19

Chemical Identifiers

Chemical Name or Material LUF 6283
Assay Percent Range >98%
Molecular Formula C8H12N2O2
Formula Weight 168.19
Synonym 5-Butyl-1H-pyrazole-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

488110

 
r & d systems
RDS488110
10mg Each for $129.05

488150

 
r & d systems
RDS488150
50mg Each for $546.19
Description & Specifications

Specifications

Formula Weight 168.19
Quantity 10mg
Chemical Name or Material LUF 6283
Assay Percent Range >98%
Molecular Formula C8H12N2O2
CAS 92933-48-7
Synonym 5-Butyl-1H-pyrazole-3-carboxylic acid
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C