missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ LRRK2 Recombinant Human Protein DFS Item

LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain.

$1,786.00 - $7,880.00

Specifications

Format Frozen
Host Species Human
Protein Tag GST Tag
Recombinant Yes
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR8604B View Documents Invitrogen™
LSPR8604B
100μg Each for $1,786.00
 
PV4874 View Documents Invitrogen™
LSPV4874
1mg Each for $7,880.00
 
Description

Description

Mutations in the LRRK2 gene have been associated with Parkinson's disease.

Specifications

Specifications

Frozen
Human
GST Tag
Yes