LRRK2 [R1441C] Human Protein, Invitrogen™ DFS Item
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain.
Manufacturer:  Life Technologies LSPV5859
 View more versions of this product
Catalog No. PV5859
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Mutations in this gene have been associated with Parkinson 's disease, including the R1441C mutation in the GTPase domain.
Specifications
Specifications
NP_940980.2
Frozen
1mg
RUO - research use only
LRRK2 R1441C
Baculovirus
Human
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
204.8kDa
Kinase proteins are shipped on dry ice and should be stored at -80°C.
Catalytic
Kinase Assay
LRRK Kinase Family
GST Tag
Documents