missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Human LRRK2 [G2019S], GST Tag Recombinant Protein
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain.
Supplier: Thermo Fisher Scientific PV4882
Description
Mutations in the LRRK2 gene have been associated with Parkinson's disease. The G2019S mutation of LRRK2 is associated with an increase in catalytic activity.
Specifications
NP_940980.2, G2019S | |
204.9 kDa | |
LRRK2 G2019S | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: TKL Group | |
Baculovirus | |
Recombinant, Catalytic, Mutant | |
Signal Transduction |
Kinase Assay | |
1mg | |
LRRK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Polysorbate 20, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |