Please login to your online account to display your discounted pricing

LP 44, Tocris

High affinity 5-HT7 agonist

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material LP 44
Assay Percent Range >98%
Molecular Formula C27H37N3OS·HCl
Formula Weight 488.13
Synonym 4-[2-(Methylthio)phenyl]-N-(1,2,3,4-tetrahydro-1-naphthalenyl)-1-piperazinehexanamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

253410

 
r & d systems
253410
10mg Each for $257.85

253450

 
r & d systems
253450
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 488.13
Quantity 10mg
Chemical Name or Material LP 44
Assay Percent Range >98%
Molecular Formula C27H37N3OS·HCl
CAS 824958-12-5
Synonym 4-[2-(Methylthio)phenyl]-N-(1,2,3,4-tetrahydro-1-naphthalenyl)-1-piperazinehexanamide hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature