Please login to your online account to display your discounted pricing

LP 20 hydrochloride, Tocris™

Selective 5-HT7 ligand

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material LP 20 hydrochloride
Assay Percent Range >98%
Molecular Formula C17H20N2O·HCl
Formula Weight 304.81
Synonym 1-(4'-Methoxy[1, 1-biphenyl]-2-yl)piperazine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

470410

 
r & d systems
470410
10mg Each for $221.01

470450

 
r & d systems
470450
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 304.81
Quantity 10mg
Chemical Name or Material LP 20 hydrochloride
Assay Percent Range >98%
Synonym 1-(4'-Methoxy[1, 1-biphenyl]-2-yl)piperazine hydrochloride
CAS 1386928-34-2
Molecular Formula C17H20N2O·HCl
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature