Please login to your online account to display your discounted pricing

LP 12 hydrochloride, Tocris

5-HT7 agonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material LP 12 hydrochloride
Assay Percent Range >99%
Molecular Formula C32H39N3O·HCl
Formula Weight 518.13
Synonym 4-(2-Diphenyl)-N-(1,2,3,4-tetrahydronaphthalen-1-yl)-1-piperazinehexanamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

292510

 
r & d systems
292510
10mg Each for $239.63

292550

 
r & d systems
292550
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 518.13
Quantity 10mg
Assay Percent Range >99%
Recommended Storage Desiccate at Room Temperature
Chemical Name or Material LP 12 hydrochloride
Synonym 4-(2-Diphenyl)-N-(1,2,3,4-tetrahydronaphthalen-1-yl)-1-piperazinehexanamide hydrochloride
CAS 1185136-22-4
Molecular Formula C32H39N3O·HCl
Safety and Handling

Desiccate at Room Temperature