missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Low-Retention Pipette Tips, Nonfilter

Increase accuracy by reducing sample retention

$144.21 - $172.61

Specifications

Filtered Non-Filtered
Color Natural
Tip Style Extended Length
For Use With (Equipment) Universal Fit
View More Specs

Products 2
Catalog Number Mfr. No. Description Volume (Metric) Tip Style Price Quantity  
Catalog Number Mfr. No. Description Volume (Metric) Tip Style Price Quantity  
02-717-132
View Documents Promotion Details
N/A Low retentionSterile, Rack 10 μL Extended Length
Pack of 960 Each for $172.61
Case of 5 Pack for $863.05
 
02-717-150
View Documents Promotion Details
N/A Low RetentionNon-sterile, Rack 1000 μL Extended Length
Pack of 1000 Each for $144.21
Case of 5 Pack for $721.05
 
Description

Description

Increase accuracy by reducing sample retention

  • Low retention surface minimizes samples loss
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
  • Natural color
Specifications

Specifications

Non-Filtered
Natural
Extended Length
Universal Fit
SDS
missing translation for 'documents'

missing translation for 'documents'