Please login to your online account to display your discounted pricing

Thermo Scientific™ Low Profile Strip Tubes without Caps


Increase the efficiency of reactions with these low-profile strip tubes for PCR and qPCR applications.

Manufacturer: thermo scientific™  AB0848

 View more versions of this product

Catalog No. AB-0848

  • / Case of 80 Each
 

Description & Specifications

Specifications

Type Strips of 12; 10 Packs of 8
Cap Type Domed
Certifications/Compliance Certified free of human DNA, RNase, DNase, and endotoxins
Color Natural
Description ABgene Low Profile Strip Tubes

The low profile design of the Thermo Scientific strip tubes without caps reduces the 'dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Low profile products are especially recommended for use with reaction volumes below 20 µL.

The low profile design of the Thermo Scientific Low Profile Strip Tubes reduces the dead space between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Thermo Scientific low profile products are especially recommended for use with reaction volumes below 20 µL.

Highlights:
  • Suitable for 0.2 mL thermal cycler blocks
  • 150 µL maximum tube capacity when capped
  • Low profile to reduce dead space and increase PCR efficiency
  • Labelled A–H end tabs
  • Ultra Clear caps are ideal for qPCR
Applications:
PCR, qPCR and Real-Time PCR

Certifications:
Certified free from human DNA, RNases, and DNases.