missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lovibond™ Solution: PAN; 0.1%
Pre-measured, unit dose reagent for water quaility testing
Supplier: Lovibond™ 530630
Description
- Specifically formulated to serve as direct replacement to Hach™ PAN Indicator Solution
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
US EPA | |
50 mL | |
Test kits, colorimeters and spectrophotometers | |
0.01 to 0.7mg/L | |
PAN Indicator Solution |
N,N-dimethylformamide 40 to 50%, Octylphenol Decaethylene 5 to 10%, distilled Water, conductivity or of similar purity 20 to 35%, non-hazardous components >5% | |
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
50 mL | |
PAN | |
1.69 oz. |