Please login to your online account to display your discounted pricing

Lovibond™ Solution: Alkaline-Cyanide


Pre-measured, unit dose reagent for water quaility testing

Manufacturer: lovibond™  ORBL530620

 View more versions of this product

Catalog No. 09-053-207

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Volume (English) 1.69 oz.
Method PAN
Product Type Alkaline Cyanide Solution
Volume (Metric) 50mL
Certifications/Compliance US EPA
Composition Sodium Cyanide 5 to 10%, Sodium Hydroxide 1 to 5%, distilled Water, conductivity or of similar purity 90 to 100%
Range 0.01 to 0.7mg/L