Please login to your online account to display your discounted pricing

Lovibond™ Solution: Alkaline-Cyanide


Pre-measured, unit dose reagent for water quaility testing

Manufacturer: lovibond™  ORBL530621

 View more versions of this product

Catalog No. 09-534-454

  • / Each
 

Description & Specifications

Specifications

Format Fluid
For Use With (Equipment) MC500, MD600 colorimeter, SP600 spectrophotometer, SC450, MD100 Manganese colorimeters and Manganese low range reagent set
pH Range 13.7
Storage Requirements 20°C
Volume (Metric) 100mL
Product Type Alkaline Cyanide Solution
For Use With (Application) Water quality testing
Volume (English) 3.38 oz.
Color Colorless