Please login to your online account to display your discounted pricing

Lovibond™ Replacement Reagents for CHECKIT™ Comparator: DEHA Solution


Manufacturer: lovibond™  ORBL461181

 View more versions of this product

Catalog No. 09-053-200

  • / Each
 

Description & Specifications

Specifications

Format Liquid
For Use With (Equipment) Colorimeter and spectrophotometer
Volume (English) 3.38 oz.
Volume (Metric) 100mL
Product Type DEHA Reagent Solution
Range 0 to 0.5mg/L
Composition Nitric Acid 2.5 to 5%
Storage Requirements 20°C
Color Yellow
For Use With (Application) Water analysis
pH Range 0.1