Please login to your online account to display your discounted pricing

Lovibond™ Replacement Reagents for CHECKIT™ Comparator: DEHA Solution


Manufacturer: lovibond™  ORBL461185

 View more versions of this product

Catalog No. 09-053-199

  • / Each
 

Description & Specifications

Specifications

Format Liquid
For Use With (Equipment) Colorimeter and spectrophotometer
pH Range 0.1
Storage Requirements 20°C
Volume (Metric) 15mL
Color Yellow
Product Type DEHA Reagent Solution
Composition Nitric Acid 2.5 to 5%
For Use With (Application) Water analysis
Volume (English) 0.5 oz.
Range 0 to 0.5mg/L