Please login to your online account to display your discounted pricing

Lovibond™ Powder Packs™ for Total Chlorine


Premeasured unit dose DPD reagents for water quality testing

Manufacturer: lovibond™  ORB530133

 View more versions of this product

Catalog No. 09-053-225

  • / Each
 

Description & Specifications

Specifications

Product Type DPD Total Chlorine Reagent
Format Powder
Range 0 to 5mg/L
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
Certifications/Compliance US EPA
Shelf Life 5 Years
Volume (Metric) 25mL
For Use With (Equipment) SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer
Volume (English) 0.84 oz.
Quantity 1000 Pack
Accessories

5 Years