Lovibond™ Total Chlorine Powder Packs™
Premeasured unit dose DPD reagents for water quality testing
Supplier: Lovibond™ 530130
Description
Premeasured unit dose DPD reagents for water quality testing
- For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
- Also suitable for use in Hach™ instruments and test kits
- Easy-to-open, sealed foil packets eliminate risk of cross-contamination
- Available for 5, 10, and 25mL sample sizes
- Extended shelf life of up to 5 years
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted DPD method
Specifications
US EPA | |
Powder | |
SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer | |
0 to 5 mg/L | |
0.84 oz. |
25 mL | |
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
100 Pack | |
DPD Total Chlorine Reagent |