Please login to your online account to display your discounted pricing

Lovibond™ Powder Packs™ for Total Chlorine


Premeasured unit dose DPD reagents for water quality testing

Manufacturer: lovibond™  ORB530123

 View more versions of this product

Catalog No. 09-053-223

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance US EPA
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer
Format Powder
Quantity 1000 Pack
Range 0 to 2mg/L
Shelf Life 5 Years
Volume (English) 0.33 oz.
Volume (Metric) 10mL
Product Type DPD Total Chlorine Reagent
Accessories

5 Years