Lovibond™ Powder Packs™: Silica HR Molybdate
Pre-measured, unit dose reagents for water quality testing
$902.46 - $1,051.83
Specifications
Product Type HR Molybdate
For Use With (Application) Water quality testing
For Use With (Equipment) Colorimeter
View More Specs
Products 2
Catalog Number Mfr. No. Range Price Quantity  
Catalog Number Mfr. No. Range Price Quantity  
09-053-752 View Documents Lovibond™
531723
0 to 100mg/L Each for $902.46
09-053-755 View Documents Lovibond™
531953
0 to 200mg/L Each for $1,051.83
Description
Description
  • Simply tear open Powder Pack™ and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open the package
  • Easily create a “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in the corners
Specifications
Specifications
HR Molybdate
Water quality testing
Colorimeter