Please login to your online account to display your discounted pricing

Powder Packs™: Silica HR Citric Acid


Pre-measured, unit dose reagents for water quality testing

Manufacturer: tintometer inc.  ORBRP194-3

 View more versions of this product

Catalog No. 09-053-754

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Water quality testing
Storage Requirements 20°C
Format Powder
For Use With (Equipment) Hach™ and other manufacturers test kits, colorimeters and spectrophotometers
pH Range 2.2
Volume (Metric) 25mL
Color White
Product Type Silica HR Citric Acid Reagent
Range 0 to 200mg/L