Please login to your online account to display your discounted pricing

Lovibond™ Powder Packs™: Silica HR Citric Acid


Pre-measured, unit dose reagents for water quality testing

Manufacturer: lovibond™  ORBRP171-3

 View more versions of this product

Catalog No. 09-053-751

  • / Each
 

Description & Specifications

Specifications

Format Powder
For Use With (Application) Water quality testing
For Use With (Equipment) Hach™ and other manufacturers test kits, colorimeters and spectrophotometers
pH Range 2.2
Storage Requirements 20°C
Volume (Metric) 10mL
Color White
Product Type Silica HR Citric Acid Reagent
Range 0 to 1.6mg/L