Lovibond™ Optical Glass Colorimetry Cells Accessories
Precision fused cuvettes and cells are available in a range of dimensions and path lengths, and are made from optical glass
$609.76 - $695.26
Specifications
For Use With (Application) Colorimetry
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With Type Price Quantity  
Catalog Number Mfr. No. For Use With Type Price Quantity  
09-053-326 Lovibond™
ORB131905
W600 Optical Glass Cells Cell Holder Each for $609.76
09-053-349 Lovibond™
ORB690584
W600 Optical Glass Cells, 33mm path length Cell Cover Pack of 5 Each for $652.80
09-053-359 Lovibond™
ORB690599
W680 Optical Glass Cells, 50mm path length Cell Cover Pack of 5 Each for $695.26
Specifications
Description
W600 Optical Glass and Borosilicate Cells
  • For use in Model Fs, PFXi/PFX195, 880, 950 and 995 instruments with non-heated samples (up to 40°C)
  • Dimensions: 21 × 40mm
W680 Optical Glass
  • For use in Comparator 2000+, Comparator 3000 with non-heated samples (up to 40°C)
  • Dimensions: 17 × 75mm
Description & Specifications
Specifications
Colorimetry