Please login to your online account to display your discounted pricing
Lovibond™ Model 611-A Aqua Tester Interchangeable Color Discs
Optically standardized; guaranteed never to fade or change color
$490.03 - $501.57
Specifications
For Use With (Equipment) 611-A Aqua Tester
Includes Necessary glassware, reagent
View More Specs
Products
Catalog Number Mfr. No. Range Price Quantity    
15-398-164B
lovibond™
ORB61110
0 to 70 color units Each for $501.57
15-398-164C
lovibond™
ORB61111
5 to 100 color units Each for $490.03
15-398-171A
lovibond™
ORB61110A
0 to 50 color units Each for $490.03
Description & Specifications
Specifications
For Use With (Equipment) 611-A Aqua Tester
Includes Necessary glassware, reagent
Nine or 10 precision standards permanently mounted in plastic disc. Discs are supplied with all the necessary glassware and reagents.