Lovibond™ Model 611-A Aqua Tester Interchangeable Color Discs
Optically standardized; guaranteed never to fade or change color
Manufacturer: Lovibond™ ORB61110
Description
Nine or 10 precision standards permanently mounted in plastic disc. Discs are supplied with all the necessary glassware and reagents.
Specifications
Interchangeable Color Disc | |
611-A Aqua Tester | |
0 to 70 color units | |
Necessary glassware, reagent |