missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lovibond™ MC500, MD600 Colorimeter Replacement Reagents: pH value

For use with MC500 and MD600 multi-parameter colorimeter

$51.72 - $51.72

Specifications

For Use With (Application) Water quality testing
Method Phenol red
Storage Requirements 20°C
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Color Format pH Range Price Quantity  
Catalog Number Mfr. No. Color Format pH Range Price Quantity  
15-398-478 Lovibond™
ORBL471040
Red Liquid 7.7 Each for $51.72
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

Water quality testing
Phenol red
20°C