Lovibond™ MD600 Colorimeter Replacement Reagents: COD
For use with MD600 multi-parameter colorimeter
Supplier: Lovibond™ TT2420727
Description
- Parameter: COD
- Method: Dichromate /H2SO4
Specifications
US EPA | |
1 | |
Water quality testing | |
150 Tests | |
20°C | |
COD Tube Test Reagent |
Yellow to Brown | |
Liquid | |
MC500 multi-parameter colorimeter | |
0 to 15,000 mg/L | |
Dichromate/H2SO4 |