Lovibond™ MC500 Colorimeter Replacement Reagents: COD
For use with MC500 multi-parameter colorimeter
Manufacturer:  Lovibond™ 2420725
 View more versions of this product
Catalog No. 15-398-516
Description
Description
  • Parameter: COD
  • Method: Dichromate /H2SO4
Specifications
Specifications
Yellow to Brown
COD Tube Test Reagent
Fluid
MC500 multi-parameter colorimeter
1
Dichromate/H2SO4
US EPA
0 to 150mg/L
Water quality testing
150.0
20°C
Documents