missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lovibond™ MD600 Colorimeter Replacement Reagents: COD

For use with MD600 multi-parameter colorimeter

$873.44 - $873.44

Specifications

Certifications/Compliance US EPA
Color Yellow to Brown
pH Range 1
Format Liquid
For Use With (Application) Water quality testing
View More Specs

Products 3
Catalog Number Mfr. No. Range Format Price Quantity  
Catalog Number Mfr. No. Range Format Price Quantity  
15-398-517
View Documents
Lovibond™
2420726
0 to 1500 mg/L Liquid
Each for $873.44
 
15-398-516
View Documents
Lovibond™
2420725
0 to 150 mg/L Liquid
Each for $873.44
 
15-398-518
View Documents
Lovibond™
TT2420727
0 to 15,000 mg/L Liquid
Each for $873.44
 
Description

Description

  • Parameter: COD
  • Method: Dichromate /H2SO4

Specifications

Specifications

US EPA
1
Water quality testing
150 Tests
Dichromate/H2SO4
Yellow to Brown
Liquid
MC500 multi-parameter colorimeter
20°C
COD Tube Test Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'