Lovibond™ MC500 Colorimeter Replacement Reagents: Chlorine
For use with MC500 multi-parameter colorimeter
Manufacturer:  Lovibond™ RS1056
 View more versions of this product
Catalog No. 15-398-481
Specifications
Specifications
Chlorine Reagent Set
Water quality testing
15mL DPD No. 1 buffer solution, 15mL DPD No. 1 reagent solution, 15mL DPD No. 3 reagent solution
DPD
0.02 to 4mg/L
MC500 multi-parameter colorimeter
50.0
Documents