Please login to your online account to display your discounted pricing
Lovibond™ MC500 Colorimeter Replacement Reagents: Chlorine
For use with MC500 multi-parameter colorimeter
$168.72 - $171.47
Specifications
Product Type Chlorine Reagent Set
For Use With (Application) Water quality testing
For Use With (Equipment) MC500 multi-parameter colorimeter
Method DPD
View More Specs
Includes:  15-398-481
  • DPD No. 1 Buffer Solution, 15mL
  • DPD No. 1 Reagent Solution, 15mL
  • DPD No. 3 Reagent Solution, 15mL
15-398-482
  • DPD No. 1 Tablets, 100 pack
  • DPD No. 3 Tablets, 100 pack
Products
Catalog Number Mfr. No. Includes No. of Tests Range Price Quantity    
15-398-481
lovibond™
RS1056
15mL DPD No. 1 buffer solution, 15mL DPD No. 1 reagent solution, 15mL DPD No. 3 reagent solution 50.0 0.02 to 4mg/L Each for $171.47
15-398-482
lovibond™
RS1060
DPD No. 1 tablets (100 pack), DPD No. 3 tablets (100 pack) 100.0 0.01 to 6mg/L Each for $168.72
Description & Specifications
Specifications
Product Type Chlorine Reagent Set
For Use With (Application) Water quality testing
For Use With (Equipment) MC500 multi-parameter colorimeter
Method DPD
  • Parameter: Chlorine
  • Method: DPD
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.