Please login to your online account to display your discounted pricing
Lovibond™ Conformance Filter Sets
Glass Standards
Manufacturer: lovibond™  ORB139370
 View more versions of this product
Catalog No. 09-053-329
  • / Each
Description & Specifications
Specifications
Certifications/Compliance ISO 9001: 2000 quality management system
For Use With PFX-195/7, PFXi-195/7 colorimeters
Quantity 5 Filters of 15, 30, 60, 85, 100 values
Material Glass
Type Honey color (Pfund equivalents) filter set
For Use With (Application) Allow quick and simple conformance check