Please login to your online account to display your discounted pricing
Lovibond™ Conformance Filter Sets
Glass Standards
Manufacturer: lovibond™  ORB139510
 View more versions of this product
Catalog No. 09-053-327
  • / Each
Description & Specifications
Specifications
For Use With PFX-195/2, PFX1-195/2, PFX-880/P, PFXi-880/P, PFX-950/P, PFXi-950/P, PFX-995/P, PFXi-995/P colorimeters
Material Glass
Certifications/Compliance ISO 9001: 2000 quality management system
For Use With (Application) Allow quick and simple conformance check
Quantity 3 Filters of 0.5, 3.5, 5 values
Type ASTM color filter set