Please login to your online account to display your discounted pricing
Lovibond™ Conformance Filter Sets
Glass Standards
Manufacturer: lovibond™  ORB341000
 View more versions of this product
Catalog No. 09-053-367
  • / Each
Description & Specifications
Specifications
Certifications/Compliance ISO 9001: 2000 quality management system
For Use With AF650 Petroleum Oils comparator
Type Lovibond™ ASTM color filter set
For Use With (Application) Allow quick and simple conformance check
Material Glass
Quantity 3 Filters of 0.5, 3.5, 5 values